Learn More
Aviva Systems Biology Corp ARSE Antibody - middle region (ARP41665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARSE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARSE. Alternate names: CDPX, CDPX1, CDPXR, MGC163310, ASEApplicationWB. Species reactivity: Human
Catalog No. 50-014-57939