Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5063 Step-In Tall Gear Bag

Catalog No. 19819761 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-761 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-761 Supplier Ergodyne™ Supplier No. 13063

Arsenal™ 5063 Step-In Tall Gear Bag is designed to hold responder gear in a ready state.

  • 1000D nylon, double coated
  • 2 in. webbing handle and shoulder straps
  • Heavy-duty #10 zippers
  • Maltese cross
  • Center opening

Specifications

Color Red
Material 1000D Double-coated Nylon
Dimensions (L x W x H) 20 x 18 x 15 in. (51 x 46 x 38 cm)
For Use With (Application) Fire Fighters, EMTs, Biochem, Hazmat
Quantity 1 Ea.
Volume (Metric) 88 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.