Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5082 SCBA Mask Bag

Catalog No. 19819766 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-766 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-766 Supplier Ergodyne™ Supplier No. 13082
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arsenal™ 5082 SCBA Mask Bag provides smart storage and protection of vital respirator masks.

  • 1000D nylon main material
  • Bottom access to the bag drops mask into users hand
  • Hook and loop closure with two web tabs to pull apart
  • Reflective strip for better visibility
  • Spring loaded steel clasp to attach to bag, belt, or other gear
  • ID window for user identification

Specifications

Color Red
Material 1000D Nylon
Dimensions (L x W x H) 8.5 x 8.5 x 14 in. (22 x 22 x 36 cm)
For Use With (Application) Firefighters, EMT, Mining
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.