Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5116 General Duty Gear Bag

Catalog No. 19819720 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-720 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-720 Supplier Ergodyne™ Supplier No. 13016
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arsenal™ 5116 General Duty Gear Bag is designed to store, organize, and easily carry everything you need for the workday or weekend.

  • Durable material withstands wear and tear
  • 1680D ballistic polyester base reinforces the bottom for ultimate durability
  • Water-resistant backing helps to repel water and stains
  • Heavy-duty webbing straps on top and side of duffel for extra reinforcement
  • Removable and adjustable padded shoulder strap for easy transportation
  • Heavy-duty zippers to secure gear inside the duffel
  • Nickel-plated Storm Drains™ added to each compartment for ventilation
  • Two large side pockets for cell phones, headphones, pens, keys, and other small parts

Specifications

Color Black
Material 600D Polyester
Dimensions (L x W x H) 23.5 x 9.5 x 12 in. (60 x 24 x 30 cm)
For Use With (Application) Contractors, Construction, Electricians, General Purpose
Quantity 1 Ea.
Size Medium
Volume (Metric) 44 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.