Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5120 Large Wheeled Gear Bag

Catalog No. 19819768 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-768 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-768 Supplier Ergodyne™ Supplier No. 13120
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arsenal™ 5120 Large Wheeled Gear Bag includes a telescoping pull handle and wheels.

  • 1200D polyester main material
  • Molded wheels for safe and easy transport
  • Three removable internal dividers for organization
  • Molded retractable pull handle
  • Padded and adjustable shoulder strap
  • Heavy-gauge nylon stitching and heavy-duty zippers
  • Material backing provides water protection

Specifications

Color Black
Material 1200D Polyester
Dimensions (L x W x H) 32.5 x 14 x 12.5 in. (83 x 35 x 32 cm)
For Use With (Application) Contractors, Trades, Construction, Electricians, General Purpose
Quantity 1 Ea.
Volume (Metric) 93 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.