Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5130 Fall Protection Gear Bag

Catalog No. 19819772 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-772 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-772 Supplier Ergodyne™ Supplier No. 13130

missing translation for 'validMainframeMsg'

Arsenal™ 5130 Fall Protection Gear Bag is designed to store fall protection equipment.

  • 600D polyester
  • Drawstring opening and barrel lock closure
  • Webbing carry strap handle
  • Grommeted drain hole
  • Flap to protect from dust-filled environments
  • Reflective stripe

Specifications

Color Black
Material 1000D Nylon
Dimensions (L x W x H) 10 x 10 x 15 in. (26 x 26 x 38 cm)
For Use With (Application) Heavy Construction, Masonry, Ironworkers
Quantity 1 Ea.
Volume (Metric) 25 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.