Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5184 Mouthbit Respirator Bag

Catalog No. 19819787 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-819-787 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-787 Supplier Ergodyne™ Supplier No. 13184
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouthbit bag makes respiratory equipment easily accessible with hook and loop attachment.

  • 420D nylon
  • Plastic buckle closure on waist strap
  • Cover flap with hook and loop closure

Specifications

Color Black
Material 420D Nylon
Model 13184
Capacity (Metric) 2 L
For Use With (Application) Construction, Carpentry, Industrial, Painting
Quantity 1 Ea.
Dimensions (L x W x H) 5 x 4 x 6.5 in. (13 x 10 x 17 cm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.