Learn More
Ergodyne™ Arsenal™ Topped Tool Pouch
Catalog No. 19819854 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Snap-hinge Closure
Snap-hinge Zipper Closure
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-819-854 Snap-hinge Closure
19-819-859 Snap-hinge Zipper Closure
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-819-854 Supplier Ergodyne™ Supplier No. 13627
Add to Cart
Add to Cart
This item is not returnable. View return policy
Houses tools, small parts and hardware
  • One-handed snap-hinge with optional zipper closure system opening offers safety and security
  • Modular buckle connection system on the back easily connects to standard tool belts and fall protection harnesses without the worker having to remove them.
  • Tool lanyards can be attached to two D-rings on the holster for additional safety and security.
  • Can be used as both a tool pouch, tool bag or bolt bag
Specifications
Product Type Snap-hinge Closure
No. per Pack 1 Ea.