Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenazo III Dye Solution (Reagent AR), Reagents

Catalog No. p-200021995
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-194 1 L
01-336-193 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-194 Supplier Reagents Holdings Llc Supplier No. C83029001C
Only null left
Add to Cart
Add to Cart

Arsenazo III Dye Solution (Reagent AR), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color Red/Purple
Volume (Metric) 1 L
Packaging Glass Bottle
Quantity 1 L
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.