Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Arsenic QC Trace Metal Standard, 1000 mg/L, 125 mL

Catalog No. 502391695 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Arsenic QC Trace Metal Standard is designed for use with ICP-OES, ICP-MS, and AA methods, featuring a HNO3 matrix.

  • Concentration of 1000 mg/L
  • Volume of 125 mL
  • Liquid formulation
  • Suitable for environmental applications
  • Designed for use with ICP-OES, ICP-MS, and AA methods
  • One 15 mL screw-cap vial yields up to 2 liters after dilution
  • Contains hazardous material requiring special freight handling

Catalog No. 50-239-1695 Supplier Waters Corp Supplier No. 013
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Arsenic QC Trace Metal Standard is designed for use with ICP-OES, ICP-MS, and AA methods, featuring a HNO3 matrix.

  • Concentration of 1000 mg/L
  • Volume of 125 mL
  • Liquid formulation
  • Suitable for environmental applications
  • Designed for use with ICP-OES, ICP-MS, and AA methods
  • One 15 mL screw-cap vial yields up to 2 liters after dilution
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.