Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic Acid Heptahydrate Sodium Salt, MP Biomedicals

Catalog No. ICN15039910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15039910 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15039910 Supplier MP Biomedicals, Inc Supplier No. 0215039910

Specifications

CAS 10048-95-0
Quantity 10 g
Molecular Formula AsH15Na2O11
MDL Number MFCD00149179
Synonym disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate
InChI Key KOLXPEJIBITWIQ-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O
IUPAC Name disodium heptahydrate hydrogen arsorate
Molecular Weight (g/mol) 312.01
PubChem CID 61460
Formula Weight 312
Chemical Name or Material Arsenic Acid Heptahydrate Sodium Salt
Show More Show Less
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.