Learn More
Hach Company Distillation Apparatus - Arsenic Set (General Purpose Set and Heater and Support Apparatus also required)
Catalog No. 501995676 Shop All Hach Company Products
Encompass
Fast set-up for sample pretreatment. Distillation sets include only EPAapproved glassware. Easy to set up because you only receive the parts you need. Illustrated, step-by-step procedures simplify distillation. Non-mercury thermometer included.
Catalog No. 50-199-5676 Supplier Hach Company Supplier No. 2265400
Fast set-up for sample pretreatment. Distillation sets include only EPAapproved glassware. Easy to set up because you only receive the parts you need. Illustrated, step-by-step procedures simplify distillation. Non-mercury thermometer included.