Learn More
Hach Company Arsenic Dual Range Test Kit
Catalog No. 5000256951 Shop All Hach Company Products
Encompass
Our dual range Arsenic Test Kits provide field measurements for arsenic in two ranges 0-250 ppb and 0-2000 ppb These kits are an essential tool for the detection of arsenic in water helping to prevent adverse health effects and environmental contamination These kits are particularly useful for monitoring arsenic levels in drinking and well water especially in areas with natural deposits or contamination from industrial activities like mining or agriculture Arsenic Test Kits are also valuable for screening groundwater for arsenic contamination In agriculture farmers employ these kits to ensure that irrigation water is free from arsenic which can accumulate in plants and enter the food supply Our kits help you maintain water safety and comply with environmental standards providing peace of mind and protection for both health and the environment Replacement reagent set product number 100735RGT
Catalog No. 50-002-56951 Supplier Hach Company Supplier No. 100735
Our dual range Arsenic Test Kits provide field measurements for arsenic in two ranges 0-250 ppb and 0-2000 ppb These kits are an essential tool for the detection of arsenic in water helping to prevent adverse health effects and environmental contamination These kits are particularly useful for monitoring arsenic levels in drinking and well water especially in areas with natural deposits or contamination from industrial activities like mining or agriculture Arsenic Test Kits are also valuable for screening groundwater for arsenic contamination In agriculture farmers employ these kits to ensure that irrigation water is free from arsenic which can accumulate in plants and enter the food supply Our kits help you maintain water safety and comply with environmental standards providing peace of mind and protection for both health and the environment Replacement reagent set product number 100735RGT