Learn More
Strem, An Ascensus Company CAS# 7784-34-1. 5g. Arsenic(III) chloride (99.999%-As) PURATREM. MFCD00014165
SDP
Catalog No. 5090115082
Encompass
CAS# 7784-34-1. 5g. Arsenic(III) chloride (99.999%-As) PURATREM. MFCD00014165. Molecular Weight: 181.27. Molecular Formula: AsCl3. Color/form: colorless liq. Strem# 93-3309. http://www.strem.com/catalog/v/93-3309/
Catalog No. 50-901-15082 Supplier Strem, An Ascensus Company Supplier No. 9333095G
CAS# 7784-34-1. 5g. Arsenic(III) chloride (99.999%-As) PURATREM. MFCD00014165. Molecular Weight: 181.27. Molecular Formula: AsCl3. Color/form: colorless liq. Strem# 93-3309. http://www.strem.com/catalog/v/93-3309/