Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Arsenic(III) Oxide

Catalog No. 2026735G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-267-35G 5 g
20-267-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 20-267-35G Supplier Sigma Aldrich Supplier No. 2026735G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Arsenic(III) oxide (ATO) is an inorganic form of arsenic that is used as a therapeutic compound for treating patients with acute promyelocytic leukemia (APL). Many drug-delivery methods are currently being developed using nanotechnology to overcome its poor bioavailability due to rapid clearance from blood.

Specifications

CAS 1327-53-3
Quantity 5 g
Molecular Formula As2O3
Linear Formula As2O3
MDL Number MFCD00003433
Synonym ATO; Diarsenic trioxide;; Arsenic trioxide; Arsenous acid
Molecular Weight (g/mol) 197.84 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.