Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) oxide, Certified Reference Material, For titrimetry, Certified by BAM, 99.5%, MilliporeSigma™ Supelco™

Catalog No. 111019530 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9530 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9530 Supplier MilliporeSigma Supelco Supplier No. 1797110G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arsenic(III) oxide is an inorganic compound. It is soluble in acids, alkalis and glycerol. It is used in pigments, ceramic, enamels and in aniline colouring industries. It has been used as decolourizing agent, wood preservative, textile mordant and froth flotation reagent component.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1327-53-3
Physical Form Neat
Linear Formula As2O3
MDL Number MFCD00003433
Synonym Arsenic trioxide; Arsenous acid
Quantity 10 g
Formula Weight 197.84
Percent Purity >99.5%
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.