Learn More
Strem, An Ascensus Company CAS# 1327-53-3. 100g. Arsenic(III) oxide, primary standard, 99.95+% (ACS). MFCD00003433
SDP
Catalog No. 5090111431
Encompass
CAS# 1327-53-3. 100g. Arsenic(III) oxide, primary standard, 99.95+% (ACS). MFCD00003433. Molecular Weight: 197.84. Molecular Formula: As2O3. Color/form: white pwdr. Strem# 33-3313. http://www.strem.com/catalog/v/33-3313/
Catalog No. 50-901-11431 Supplier Strem, An Ascensus Company Supplier No. 333313100G
CAS# 1327-53-3. 100g. Arsenic(III) oxide, primary standard, 99.95+% (ACS). MFCD00003433. Molecular Weight: 197.84. Molecular Formula: As2O3. Color/form: white pwdr. Strem# 33-3313. http://www.strem.com/catalog/v/33-3313/