Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) telluride, 99.999% (metals basis)

Catalog No. AA4170909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4170909 10 g
AA4170918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4170909 Supplier Thermo Scientific Chemicals Supplier No. 04170909
Add to Cart

CAS: 12044-54-1 | As2Te3 | 532.643 g/mol

Vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12044-54-1
Molecular Formula As2Te3
Molecular Weight (g/mol) 532.643
MDL Number MFCD00014167
InChI Key GTIUFDICMGTSPM-UHFFFAOYSA-N
Synonym arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
PubChem CID 15770922
SMILES [As](=[Te])[Te][As]=[Te]

Specifications

Melting Point 381°C
Physical Form Pieces
Quantity 10 g
UN Number UN1557
Formula Weight 532.64
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 6.5
Density 6.5
Chemical Name or Material Arsenic(III) telluride
Hazard Category H301+H331-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 234-955-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.