Learn More
Aqua Solutions Arsenic / Manganese 10 PPM
SDP
Catalog No. NC1746675
Encompass_Preferred
Composition: 3.035% Nitric Acid (7697-37-2), 0.00436% Manganese Acetate Tetrahydrate (6156-78-1), 0.000979% arsenic (7440-38-2), 96.9598% Water (7732-18-5)
Catalog No. NC1746675 Supplier Aqua Solutions Supplier No. HON138
Composition: 3.035% Nitric Acid (7697-37-2), 0.00436% Manganese Acetate Tetrahydrate (6156-78-1), 0.000979% arsenic (7440-38-2), 96.9598% Water (7732-18-5)