Learn More
Hach Company Arsenic Standard Solution, 1000 mg/L as As, 100 mL
Supplier:  Hach Company 1457142
Encompass
For accuracy checks in arsenic determinations. (NaOH matrix)
Catalog No. 50-199-5077