Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(V) Oxide 99.999%, Thermo Scientific™

Catalog No. AC192500250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC192500250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC192500250 Supplier Thermo Scientific Chemicals Supplier No. 192500250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-28-2
Molecular Formula As2O5
Molecular Weight (g/mol) 229.838
InChI Key COHDHYZHOPQOFD-UHFFFAOYSA-N
Synonym arsenic pentoxide, arsenic v oxide, arsenic anhydride, arsenic v pentoxide, arsenic pentaoxide, arsenic oxide, diarsenic pentoxide, diarsenic pentaoxide, arsenic 5+ oxide, arsenic acid anhydride
PubChem CID 14771
SMILES O=[As](=O)O[As](=O)=O

Specifications

Physical Form Pieces
Quantity 25 g
Linear Formula As2O5
Merck Index 15, 789
Formula Weight 229.84
Chemical Name or Material Arsenic(V) oxide, 99.999%
Hazard Category Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.