Learn More
Cayman Chemical ArsenobetaIn 50mg
SDP
Catalog No. 502947907
Encompass_Preferred
An organoarsenical and compatible solute; protective against B. subtilis cell death induced by high osmolarity or extreme temperatures at 1 mM; non-toxic to mice (LD50 = 10 g/kg)
Catalog No. 50-294-7907 Supplier Cayman Chemical Supplier No. 3451550 MG
An organoarsenical and compatible solute; protective against B. subtilis cell death induced by high osmolarity or extreme temperatures at 1 mM; non-toxic to mice (LD50 = 10 g/kg)