Learn More
Aviva Systems Biology Corp Arsg Antibody - C-terminal region (ARP55122_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55122_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Arsg. It was validated on Western Blot. . Alternate names: 6330406P08Rik, AI846872, MGC90567ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-007