Learn More
Chemglass Life Sciences Arsine Generator, Improved, Complete, 24/40 Joints
SDP
Catalog No. 50872930
Encompass_Preferred
Arsine Generator, Improved, Complete, 24/40 Joints. For arsine analysis using the diethyldithiocarbamate (SDDC) colorimetric method. Design also meets specifications of ACS/USP, EPA and can be used with APHA Standard 104A. Apparatus is compact, requires less set-up time and eliminates the ball and socket connection normally used. Complete item consists of a 125mL Erlenmeyer flask and scrubber tube both having 24/40 standard taper joints. Also supplied complete with one CG-145-05, size 24 Keck clip. Glass beads may be added to the absorber tube to enhance the gas mixing.
Catalog No. 50-872-930 Supplier Chemglass Life Sciences Supplier No. CG109001
Arsine Generator, Improved, Complete, 24/40 Joints. For arsine analysis using the diethyldithiocarbamate (SDDC) colorimetric method. Design also meets specifications of ACS/USP, EPA and can be used with APHA Standard 104A. Apparatus is compact, requires less set-up time and eliminates the ball and socket connection normally used. Complete item consists of a 125mL Erlenmeyer flask and scrubber tube both having 24/40 standard taper joints. Also supplied complete with one CG-145-05, size 24 Keck clip. Glass beads may be added to the absorber tube to enhance the gas mixing.