Learn More
Wilmad Labglass ARSINE GENERATR IMPRD COMPLETE
Catalog No. 50803396
Encompass_Preferred
Arsine Generator, Improved, Complete. For arsine analysis using the diethyldithocarbamate colorimetric method. Design of apparatus requires less space and is easier to clean than standard ball and socket design.
Catalog No. 50-803-396 Supplier Wilmad Labglass Supplier No. LG6962100
Arsine Generator, Improved, Complete. For arsine analysis using the diethyldithocarbamate colorimetric method. Design of apparatus requires less space and is easier to clean than standard ball and socket design.