Learn More
Research Products International Corp Art 1000 Reach Tip, Filtered, 1000ul, Sterile, 100 Tips per Tray, 8 Trays per Case
SDP
Catalog No. 50998503
Encompass
Art 1000 Reach Tip features extended tip length and prevents pipettor shaft from entering the sample vessel. Ideal tip for use with 15ml centrifuge tubes. SoftFit design reduces hand fatigue allowing tip to fit easily on pipettor. Tips with filter feature patented self-sealing barrier that prevents aerosols and liquids from passing into pipettor and protects against carry over contamination. Made of polypropylene and available presterilized in convenient 8 racks of 100 per case.
Catalog No. 50-998-503 Supplier Research Products International Corp Supplier No. 141220
Art 1000 Reach Tip features extended tip length and prevents pipettor shaft from entering the sample vessel. Ideal tip for use with 15ml centrifuge tubes. SoftFit design reduces hand fatigue allowing tip to fit easily on pipettor. Tips with filter feature patented self-sealing barrier that prevents aerosols and liquids from passing into pipettor and protects against carry over contamination. Made of polypropylene and available presterilized in convenient 8 racks of 100 per case.