Learn More
Research Products International Corp ART 100E Aerosol Resistant Tips for P-100, Capacity 100ul, 96 per Rack, 960 per Case
SDP
Catalog No. 50476634
Encompass
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable. *The PCR process is covered by patents owned by Hoffman-LaRoche.
Fits all major brands.
Catalog No. 50-476-634 Supplier Research Products International Corp Supplier No. 141046
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable. *The PCR process is covered by patents owned by Hoffman-LaRoche.
Fits all major brands.