Learn More
Research Products International Corp Art 10 Aerosol Resistant Tips, 0.5-10ul, 96 per Rack, 960 per Case
SDP
Catalog No. 50476633
Encompass
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.
Positive sealing barrier
Prevent pipettor contamination
Ideal for DNA amplification RNase, DNase, pyrogen free.
Catalog No. 50-476-633 Supplier Research Products International Corp Supplier No. 141044
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.
Positive sealing barrier
Prevent pipettor contamination
Ideal for DNA amplification RNase, DNase, pyrogen free.