Learn More
Research Products International Corp Art Aerosol Resistant Tips, 20ul Volume, Sterile, 96 per Rack, 960 per Case
SDP
Catalog No. 50550163
Encompass
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.
Positive sealing barrier
Prevent pipettor contamination
Ideal for DNA amplication RNase, DNase, pyrogen free
Catalog No. 50-550-163 Supplier Research Products International Corp Supplier No. 141069
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.
Positive sealing barrier
Prevent pipettor contamination
Ideal for DNA amplication RNase, DNase, pyrogen free