Learn More
Thomas Scientific ART 200 Non-Filtered Tip, Hinged Racked, Sterile, Low Retention, 200 uL Pipette Tips, 960/pk
SDP
Catalog No. NC1935925
Encompass
Access tips with one hand using Thermo Scientific ART Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.
Catalog No. NC1935925 Supplier Thomas Scientific Supplier No. 1199Z84
Access tips with one hand using Thermo Scientific ART Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.