Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips
GREENER_CHOICE

Catalog No. 21402482 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
1000 μL
Model:
ART 10
ART 10 REACH
ART 1000
ART 100E
ART 10F
ART 200
ART 20P
Tip Style:
Ergonomic, Low Retention
Extended Length, Low Retention, Micro Point
Low Retention, Micro Point
Fit for Purpose:
Sterile and PCR Ready
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model Tip Style Fit for Purpose
21-402-482 10 μL ART 10 REACH Extended Length, Low Retention, Micro Point Sterile and PCR Ready
21-402-5E007 10 μL ART 10 Low Retention, Micro Point Sterile and PCR Ready
21-402-481 10 μL ART 10F Low Retention, Micro Point Sterile and PCR Ready
21-402-484 20 μL ART 20P Low Retention, Micro Point Sterile and PCR Ready
21-402-485 100 μL ART 100E Low Retention, Micro Point -
21-402-486 200 μL ART 200 Low Retention, Micro Point Sterile and PCR Ready
21-402-487 1000 μL ART 1000 Ergonomic, Low Retention Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 21-402-482 Supplier Thermo Scientific™ Supplier No. 214005
  • $344.00
    $217.65 / Pack of 960

    Save $126.35

    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,509.00
    $954.65 / Case of 5 PK

    Save $554.35

    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate sample retention with these specially designed pipette tips.

Get Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips, which are specifically designed for eliminating sample retention within the pipette tip.

  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Tips seal when exposed to potential contaminants
  • Certified RNase, DNase, ATP, Endotoxin, and Pyrogen Free
TRUSTED_SUSTAINABILITY

Specifications

Description ART 10 REACH Low retention, Racked, Sterile
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Clear
Model ART 10 REACH
Length (English) 1.71 in.
Length (Metric) 4.33cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Rack
Tip Style Extended Length, Low Retention, Micro Point
Packaging Rack, 10 x 96
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.