Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack
GREENER_CHOICE

Catalog No. 212364
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 μL
100 μL
1000 μL
20 μL
200 μL
300 μL
Model:
ART 10
ART 100
ART 1000E
ART 100E
ART 10F
ART 20
ART 200
ART 300
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
21-236-4 20 μL ART 20
21-236-27 300 μL ART 300
21-236-8 100 μL ART 100E
21-236-1 200 μL ART 200
21-236-7 100 μL ART 100
21-236-2A 1000 μL ART 1000E
21-236-13 0.5 to 10 μL ART 10F
21-236-5 10 μL ART 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 21-236-4 Supplier Thermo Scientific™ Supplier No. 2149P
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform applications accurately and consistently with Barrier Pipette Tips in Lift-off Lid Racks, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier pipette tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

TRUSTED_SUSTAINABILITY

Specifications

Description ART 20, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric) 20 μL
Volume (Metric) Dispensing 20μL
Filtered Filtered
Sterility Sterile
Autoclavable Not Autoclavable
Model ART 20
Length (English) 2 in.
Length (Metric) 5.07 cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Lift-off Lid Rack
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Product Line ART
Material Polypropylene
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.