Learn More
Thermo Scientific™ ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks

Description
Unlike standard length barrier tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks self-seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. Available in MicroPoint™, low retention and offer a universal fit design.
Features:
- Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
- Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package
- Stackable hinged racks save space in your lab
- Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin
Recommended for:
- Liquid Handling
- Molecular Biology
- PCR
- Biotechnology
- Front latch closes and locks, ensuring safe and secure storage throughout
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
- Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Lid rim prevents racks from sliding around when stacked and carted around a lab
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Volume (Metric) Dispensing | 200 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Natural |
| Autoclavable | Not Autoclavable |
| Model | ART 200 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit |
| Length (English) | 2.32 in. |
| Length (Metric) | 5.88 cm |
| Certifications/Compliance | ISO 9001:2015 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.