Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
GREENER_CHOICE

Catalog No. 02681576 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
1000 μL
Tip Style:
Low Retention
Standard
Model:
ART 1000 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
ART 20 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
ART 20 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
ART 200 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
ART 200 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style Model
02-681-576 200 μL Standard ART 200 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
02-681-541 200 μL Low Retention ART 200 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
02-681-546 20 μL Low Retention ART 20 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
02-681-575 1000 μL Standard ART 1000 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
02-681-577 20 μL Standard ART 20 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-681-576 Supplier Thermo Scientific™ Supplier No. 2069RCHR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The pipette tip and rack mail-back program provides a simple and effective solution for recycling PP#5 single use plastics. The TipCycle Kit included with your case of pipette tips to collect used pipette tips and empty racks. Complete waiver, attach pre-paid shipping label and return for recycling.

Unlike standard length barrier tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks self-seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. Available in MicroPoint™, low retention and offer a universal fit design.

Features:

  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package
  • Stackable hinged racks save space in your lab
  • Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
  • Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Lid rim prevents racks from sliding around when stacked and carted around a lab
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Volume (Metric) Dispensing 200 μL
Filtered Filtered
Sterility Sterile
Color Natural
Rack Color Natural
Autoclavable Not Autoclavable
Model ART 200 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
Length (English) 2.32 in.
Length (Metric) 5.88 cm
Certifications/Compliance ISO 9001:2015
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
For Use With (Application) Liquid Handling, Molecular Biology, PCR
Sterilization Method Gamma Irradiated
Format Hinged Rack
Tip Style Standard
Packaging Hinged Rack
Quantity 4800/Cs.
Product Type TipCycle, Pipette Tip and Rack Recycling Kit
Includes 1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
ACT Label No
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line ART
Material Polypropylene
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.