Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART 1000 REACH RACKED barrier sterile pipette tips
GREENER_CHOICE

Catalog No. 2140303 Shop All Thermo Scientific Products
Change view
Click to view available options
ART Barrier Pipette tips 1000 REACH; 1000μL MicroPoint, extended length, sterile tip
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
21-403-03 ART Barrier Pipette tips 1000 REACH; 1000μL MicroPoint, extended length, sterile tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-403-03 Supplier Thermo Scientific™ Supplier No. 2079
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetting for a variety of applications:

Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

TRUSTED_SUSTAINABILITY

Specifications

Description ART 1000 REACH, Filtered, Sterile, Rack
Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Model ART 1000 REACH
Length (English) 4.66 in.
Length (Metric) 11.85cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Rack
Tip Style Extended Length
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.