Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Specialty Pipette tips
GREENER_CHOICE

Catalog No. p-4525528 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
10 μL
1000 μL
200 μL
50 μL
Model:
ART 10 REACH
ART 1000
ART 200U
ART 20E
ART 50U
Tip Style:
Ergonomic
Extended Length
Micro Point
Fit for Purpose:
Sterile and PCR Ready
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model Tip Style Fit for Purpose
21-402-185 200 μL ART 200U Ergonomic Sterile and PCR Ready
21-236-86 50 μL ART 50U Ergonomic Sterile and PCR Ready
21-236-85 1000 μL ART 1000 Ergonomic Sterile and PCR Ready
21-236-35 10 μL ART 10 REACH Extended Length Sterile and PCR Ready
21-402-161 0.5 to 20 μL ART 20E Micro Point Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 21-402-185 Supplier Thermo Scientific™ Supplier No. 2770
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to Aid in Ppipetteting for a Variety of Applications:

Extended length: to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading: for loading acrylamide or agarose gels

Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials

Solvent safe: for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

TRUSTED_SUSTAINABILITY

Specifications

Description ART 200U, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric) 200 μL
Filtered Filtered
Sterility Sterile
Autoclavable Not Autoclavable
Model ART 200U
Length (English) 2.06 in.
Length (Metric) 5.2cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Lift-off Lid Rack
Tip Style Ergonomic
For Use With (Equipment) Gilson Pipetman, Thermo Scientific Finnpipette Focus
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.