Learn More
Diversified Spaces™ Art/Drafting Table
SCI_ED
Catalog No. S42677 Shop All Diversified Spaces Products
Change view
Click to view available options
Material:
Maple
Catalog No. Material
S42677 Maple
1 options
Catalog No. S42677 Supplier Diversified Spaces™ Supplier No. LT4424M
Add to Cart
Add to Cart
Constructed of a maple hard wood.
  • Constructed of maple hard wood frame and plastic laminate almond colored top
  • Features shatter-resistant glass inset for maximum diffusion of light with minimum glare and no hot spots
  • Top adjusts from 0-30 degrees and is equipped with soft-close feature and pencil stop
  • Includes 120 volt electrical cord and four energy efficient fluorescent tubes
  • Coated with chemical resistant, earth-friendly UV finish
Certifications
MAS Certified Green
Compliance
SEFA
Warranty and Services
Limited Lifetime
Specifications
Product Type Drawing Table
Color Maple
Dimensions (D x W x H) 60.9 x 111.7 x 100.9 cm (24 x 44 x 39.75 in.)
Finish Chemical resistant, earth-friendly UV
Material Maple