Learn More
Research Products International Corp Art Gel 20P Aerosol Resistant Tips, 0 - 20ul Volume Range, 96 per Rack, 960 per Case
SDP
Catalog No. 50212198
Encompass
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.
Positive sealing barrier
Prevent pipettor contamination
Ideal for DNA amplication RNase, DNase, pyrogen free
Catalog No. 50-212-198 Supplier Research Products International Corp Supplier No. 141071
ART tips are made of polypropylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.
Positive sealing barrier
Prevent pipettor contamination
Ideal for DNA amplication RNase, DNase, pyrogen free