Learn More
Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks
GREENER_CHOICE
Catalog No. 02682270 Shop All Thermo Scientific Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
02-682-270 Small
02-682-271 Medium
02-682-272 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-682-270 Supplier Thermo Scientific™ Supplier No. 2004ER
Add to Cart
Add to Cart
Save valuable space in the lab with these stackable pipette tip hinged racks.
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks. ART barrier tips provide 100% security against aerosols and liquid contamination. Front latch closes and locks, ensuring safe and secure storage throughout.
Features:
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Certifications:
  • Nonsterile products certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
  • Sterile products certified to be free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
Shape Square
Stackable Yes
Sterility Non-sterile
Size Small
Product Type Empty Hinged Rack