Learn More
Research Products International Corp Art Reach Filter Tips, 0.5-10.0ul, Sterile, 960 per Case
SDP
Catalog No. 50998500
Encompass
ART tips are made of polyproplylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.
Catalog No. 50-998-500 Supplier Research Products International Corp Supplier No. 141093
ART tips are made of polyproplylene and are pre-sterilized in convenient racks. ART tips are not autoclavable.
The PCR process is covered by patents owned by Hoffman-LaRoche. Purchases of these products does not include a license to do PCR.