Learn More
Pickering Laboratories Inc Artificial Saliva, Fusayama/Mayer, Not Stabilized, each (200 mL/bottle)
SDP
Catalog No. NC0680078
Encompass_Preferred
This ready-to-use solution closely resembles themineral composition of natural saliva and can be usedfor testing a wide variety of products, including dentalmetal alloys. This formulation is at pH 4.9 and shouldbe stored refrigerated.
Catalog No. NC0680078 Supplier Pickering Laboratories Inc Supplier No. 17000301
This ready-to-use solution closely resembles themineral composition of natural saliva and can be usedfor testing a wide variety of products, including dentalmetal alloys. This formulation is at pH 4.9 and shouldbe stored refrigerated.