Learn More
Aviva Systems Biology Corp ART1 Antibody - C-terminal region (ARP60195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ART1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ART1. Alternate names: ART2, CD296, MGC133217, RT6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-972