Learn More
Andwin Scientific ART10 PIPET TIP 0.1-10UL (960)
SDP
Catalog No. NC9548194
Encompass_Preferred
The ART� 10 Self-Sealing Barrier Pipet Tips feature a patented, self-sealing barrier that allow complete blocking of the passage of aqueous based liquids and aerosols from the sample to the pipettor. This design provides protection to the pipetting device from any viable infection that could carry over and negatively affect future samples. This product is ideal for crucial applications, such as PCR, sequencing, loading gels, handling infectious samples and pipetting radioactive isotopes. The tips are all free of RNase, DNase, DNA, and pyrogens. It is sold in a pack of 960.
Catalog No. NC9548194 Supplier Andwin Scientific Supplier No. P2139
The ART� 10 Self-Sealing Barrier Pipet Tips feature a patented, self-sealing barrier that allow complete blocking of the passage of aqueous based liquids and aerosols from the sample to the pipettor. This design provides protection to the pipetting device from any viable infection that could carry over and negatively affect future samples. This product is ideal for crucial applications, such as PCR, sequencing, loading gels, handling infectious samples and pipetting radioactive isotopes. The tips are all free of RNase, DNase, DNA, and pyrogens. It is sold in a pack of 960.