Learn More
Aviva Systems Biology Corp ART3 Antibody - middle region (ARP45062_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45062_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ART3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ART3. Alternate names: ARTC3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58965