Learn More
Selleck Chemical LLC ART558 5MG
SDP
Catalog No. NC2332045
Encompass_Preferred
ART558 is a potent, selective, low molecular weight, allosteric inhibitor of DNA Pol? with an IC50 of 7.9 nM.CAS No. 2603528-97-6 *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC2332045 Supplier Selleck Chemical LLC Supplier No. S99365MG
ART558 is a potent, selective, low molecular weight, allosteric inhibitor of DNA Pol? with an IC50 of 7.9 nM.CAS No. 2603528-97-6 *For Research & Development use only.  Product is not intended for drug, household, or human consumption.