Learn More
TARGETMOL CHEMICALS INC ART558 5MG
SDP
Catalog No. 502579012
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ART558 is a potent selective low molecular weight allosteric DNA polymerase activity of Pol(theta) inhibitor (IC50=7.9 nM). ART558 can be used for the research of cancer. purity: 99%
Catalog No. 50-257-9012 Supplier TARGETMOL CHEMICALS INC Supplier No. T92755MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ART558 is a potent selective low molecular weight allosteric DNA polymerase activity of Pol(theta) inhibitor (IC50=7.9 nM). ART558 can be used for the research of cancer. purity: 99%