Learn More
Medchemexpress LLC HY-135578 5mg Medchemexpress, Artelinic acid CAS:120020-26-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1355785MG
Encompass_Preferred
Medchemexpress, HY-135578 5mg Artelinic acid CAS:120020-26-0 Artelinic acid, a derivative of Artemisinin, is an antimalarial drug for the treatment of multidrug resistant strains of Plasmodium falciparum. Artelinic acid can be administered by various routes of administration, including intravenous, intramuscular and oral routes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1391