Learn More
Medchemexpress LLC Artemether-d3 | 93787-85-0 | 301.39 | 1 MG
SDP
Catalog No. 5000389127
Encompass_Preferred
Artemether-d3 is a deuterium-labeled version of Artemether, an antimalarial drug used to treat resistant strains of falciparum malaria. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is largely used as tracers for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Application as a tracer.
  • Internal standard for quantitative analysis.
  • High purity.
  • Stable isotope.
Catalog No. 50-003-89127 Supplier Medchemexpress LLC Supplier No. HYN0402S1MG
Artemether-d3 is a deuterium-labeled version of Artemether, an antimalarial drug used to treat resistant strains of falciparum malaria. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is largely used as tracers for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Application as a tracer.
  • Internal standard for quantitative analysis.
  • High purity.
  • Stable isotope.