Learn More
Apexbio Technology LLC Artemether (SM-224) 71963-77-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093175
Encompass_Preferred
Artemether (SM-224 CAS 71963-77-4) is a small-molecule inhibitor designed to inhibit the proliferation of Plasmodium species and impair the viability of Schistosoma japonicum Artemether exerts its biological activity primarily through metabolism in parasites generating reactive oxygen species that damage parasite biomolecules In in vitro studies Artemether demonstrates inhibitory activity against Schistosoma mansoni with IC50 values ranging approximately from 30 50 nM Based on these pharmacological properties Artemether holds research potential in antimalarial drug development and the study of parasite infection mechanisms
Catalog No. 50-000-93175 Supplier Apexbio Technology LLC Supplier No. A40145.1
Artemether (SM-224 CAS 71963-77-4) is a small-molecule inhibitor designed to inhibit the proliferation of Plasmodium species and impair the viability of Schistosoma japonicum Artemether exerts its biological activity primarily through metabolism in parasites generating reactive oxygen species that damage parasite biomolecules In in vitro studies Artemether demonstrates inhibitory activity against Schistosoma mansoni with IC50 values ranging approximately from 30 50 nM Based on these pharmacological properties Artemether holds research potential in antimalarial drug development and the study of parasite infection mechanisms