Learn More
TARGETMOL CHEMICALS INC Artemisic acid 100MG
SDP
Catalog No. 502876657
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 1. Artemisic acid ((+)-Artemisinic Acid) shows antibacterial activity. 2. Artemisinic acid is a major precursor of artemisinin (an antimalaric compound), isolated as the active principles of the medicinal plant Artemisia annua L. Purity 99.05%
Catalog No. 50-287-6657 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S1452100MG
Add to Cart
Add to Cart
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 1. Artemisic acid ((+)-Artemisinic Acid) shows antibacterial activity. 2. Artemisinic acid is a major precursor of artemisinin (an antimalaric compound), isolated as the active principles of the medicinal plant Artemisia annua L. Purity 99.05%